Assay Details
| Gene Symbol: Pvrl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012064.1 | NP_001012064.1 | 8 - 9 | 1532 | 61 | |
| GenBank mRNA | BC090075.1 | - | 8 - 9 | 1532 | 7189358 | 61 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
acrosome assembly
positive regulation of natural killer cell mediated cytotoxicity directed against tumor cell target
positive regulation of immunoglobulin mediated immune response
cytoskeleton organization
homophilic cell adhesion via plasma membrane adhesion molecules
spermatid development
spermatid nucleus differentiation
biological_process
fertilization
fusion of virus membrane with host plasma membrane
sperm mitochondrion organization
cell part morphogenesis
positive regulation of mast cell activation
susceptibility to natural killer cell mediated cytotoxicity
adhesion of symbiont to host
cilium organization
positive regulation of natural killer cell mediated cytotoxicity
coreceptor-mediated virion attachment to host cell
establishment of mitochondrion localization
susceptibility to T cell mediated cytotoxicity
molecular_function
identical protein binding
protein homodimerization activity
cell adhesion molecule binding
acrosome assembly
positive regulation of natural killer cell mediated cytotoxicity directed against tumor cell target
positive regulation of immunoglobulin mediated immune response
cytoskeleton organization
homophilic cell adhesion via plasma membrane adhesion molecules
spermatid development
spermatid nucleus differentiation
biological_process
fertilization
fusion of virus membrane with host plasma membrane
sperm mitochondrion organization
cell part morphogenesis
positive regulation of mast cell activation
susceptibility to natural killer cell mediated cytotoxicity
adhesion of symbiont to host
cilium organization
positive regulation of natural killer cell mediated cytotoxicity
coreceptor-mediated virion attachment to host cell
establishment of mitochondrion localization
susceptibility to T cell mediated cytotoxicity
molecular_function
identical protein binding
protein homodimerization activity
cell adhesion molecule binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P290591 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672585 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830134 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567188 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948784 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496442 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461914 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17680 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385316 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171485 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281416 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555381 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309913 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940677 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955496 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897813 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971385 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72807 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144218 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P290591 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P290591 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P290591 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P290591 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 29181 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 24660 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 15533 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 56146 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10719556 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10719553 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10719559 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10719558 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10719551 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10719555 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10719557 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10719560 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10719550 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10719552 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10719549 | - | - |
| Rat Genome 230 2.0 Array | 1389681_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1375216_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1389681_at | - | - |
| Rat Genome 230 2.0 Array | 1389899_at | - | - |
| Rat Genome 230 2.0 Array | 1375216_at | - | - |
| Rat Genome 230 2.0 Array | 1389899_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1389899_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1375216_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1389681_at | - | - |
| Affymetrix HT Rat Focus Array | 1389899_at | - | - |
| Affymetrix HT Rat Focus Array | 1389681_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1375216_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100039061.rn.1 | TC0100004937.rn.1 | RTA TC |
| Rat Expression Set 230 | 1389899_at | - | - |
| Rat Expression Set 230 | 1375216_at | - | - |
| Rat Expression Set 230 | 1389899_PM_at | - | - |
| Rat Expression Set 230 | 1389681_PM_at | - | - |
| Rat Expression Set 230 | 1375216_PM_at | - | - |
| Rat Expression Set 230 | 1389681_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10719549 | - | - |
| Rat Genome U34 Set | rc_AA900198_at | - | - |
| Rat Genome U34 Set | rc_AA850909_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20876457 | - | - |
| ABI Rat Genome Survey Microarray | 21722948 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1375236 | - | - |
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376242 | - | - |

Pvrl2
Genomic Map