Assay Details
Gene Symbol: Gapdhs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023964.1 | NP_076454.1 | 2 - 3 | 324 | 80 | |
XM_006228790.2 | XP_006228852.1 | 2 - 3 | 694 | 80 | ||
XM_006228791.2 | XP_006228853.1 | 2 - 3 | 555 | 80 | ||
GenBank mRNA | AJ297631.1 | - | 2 - 3 | 324 | 80 | |
BC161847.1 | - | 2 - 3 | 346 | 7112898 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
