Assay Details
| Gene Symbol: Med25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170426.1 | NP_001163897.1 | 3 - 4 | 354 | 99 | |
| XM_006229007.2 | XP_006229069.1 | 3 - 4 | 607 | 99 | ||
| XM_006229008.2 | XP_006229070.1 | 3 - 4 | 607 | 99 | ||
| XM_006229009.2 | XP_006229071.1 | 3 - 4 | 608 | 99 | ||
| XM_006229010.2 | XP_006229072.1 | 3 - 4 | 610 | 99 | ||
| XM_006229011.2 | XP_006229073.1 | 3 - 4 | 605 | 99 | ||
| XM_006229012.2 | XP_006229074.1 | 3 - 4 | 606 | 99 | ||
| XM_006229013.2 | XP_006229075.1 | 2 - 3 | 216 | 99 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the XM_006229013.2 transcripts.
Set Membership: |
|

Med25
Genomic Map