Assay Details
| Gene Symbol: Psenen | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008764.2 | NP_001008764.1 | 2 - 3 | 172 | 65 | |
| XM_006228770.2 | XP_006228832.1 | 2 - 3 | 224 | 65 | ||
| GenBank mRNA | AY539891.1 | - | 1 - 2 | 63 | 65 | |
| BC100104.1 | - | 2 - 3 | 172 | 7377803 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Psenen
Genomic Map