Assay Details
| Gene Symbol: Bach1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107113.1 | NP_001100583.1 | 1 - 2 | 83 | 99 | |
| XM_003751009.3 | XP_003751057.2 | 1 - 2 | 128 | 99 | ||
| XM_006248018.2 | XP_006248080.1 | 1 - 2 | 128 | 99 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001107113.1, XM_003751009.3, XM_006248018.2 transcripts.
Set Membership: |
|

Bach1
Genomic Map