Assay Details
Gene Symbol: Fsd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191627.1 | NP_001178556.1 | 3 - 4 | 955 | 69 | |
XM_006229440.2 | XP_006229502.1 | 4 - 5 | 1159 | 69 | ||
XM_008759537.1 | XP_008757759.1 | 4 - 5 | 2376 | 69 | ||
XM_008759538.1 | XP_008757760.1 | 5 - 6 | 1549 | 69 | ||
GenBank mRNA | FQ223649.1 | - | 4 - 5 | 1107 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
