Assay Details
| Gene Symbol: Klhl25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039006.1 | NP_001034095.1 | 1 - 2 | 94 | 98 | |
| GenBank mRNA | BC099111.1 | - | 1 - 2 | 94 | 7461562 | 98 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC099111.1,NM_001039006.1 transcripts.
Set Membership: |
|

Klhl25
Genomic Map