Assay Details
| Gene Symbol: Unc45a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037647.1 | NP_001032736.1 | 4 - 5 | 547 | 119 | |
| XM_006229432.2 | XP_006229494.1 | 4 - 5 | 498 | 119 | ||
| GenBank mRNA | BC107919.1 | - | 4 - 5 | 547 | 7457637 | 119 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P1027602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245226 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491810 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228804 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15215 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40909 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203860 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26742 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1027602 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1027602 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P1027602 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1027602 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 65665 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10723050 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723052 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723060 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723057 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723058 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723045 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723056 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723055 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723044 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723049 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723046 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723061 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723059 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723053 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723062 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723054 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723063 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723043 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723048 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723051 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723047 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10723039 | - | - |
| Rat Genome 230 2.0 Array | 1372679_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1372679_at | - | - |
| Affymetrix HT Rat Focus Array | 1372679_at | - | - |
| Affymetrix HT Rat Focus Array | 1372679_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100046735.rn.1 | TC0100005877.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100046738.rn.1 | TC0100005877.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100046740.rn.1 | TC0100005877.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100046750.rn.1 | TC0100005877.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100046739.rn.1 | TC0100005877.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100046741.rn.1 | TC0100005877.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100046744.rn.1 | TC0100005877.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100046733.rn.1 | TC0100005877.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100046737.rn.1 | TC0100005877.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100046742.rn.1 | TC0100005877.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100046743.rn.1 | TC0100005877.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100046749.rn.1 | TC0100005877.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100046736.rn.1 | TC0100005877.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100046732.rn.1 | TC0100005877.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100046745.rn.1 | TC0100005877.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100046746.rn.1 | TC0100005877.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100046747.rn.1 | TC0100005877.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100046751.rn.1 | TC0100005877.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100046734.rn.1 | TC0100005877.rn.1 | RTA TC |
| Rat Expression Set 230 | 1372679_PM_at | - | - |
| Rat Expression Set 230 | 1372679_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10723039 | - | - |
| Rat Genome U34 Set | rc_AI179818_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21801361 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357071 | - | - |

Unc45a
Genomic Map