Assay Details
| Gene Symbol: Mrps11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109148.1 | NP_001102618.1 | 2 - 3 | 246 | 85 | |
| XM_006229400.1 | XP_006229462.1 | 3 - 4 | 201 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mrps11
Genomic Map