Assay Details
| Gene Symbol: Leprotl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013188.2 | NP_001013206.1 | 1 - 2 | 71 | 88 | |
| GenBank mRNA | BC058504.1 | - | 1 - 2 | 75 | 6887991 | 88 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC058504.1, NM_001013188.2 transcripts.
Set Membership: |
|

Leprotl1
Genomic Map