Assay Details
| Gene Symbol: Edrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107557.1 | NP_001101027.1 | 1 - 2 | 233 | 79 | |
| XM_006230436.2 | XP_006230498.1 | 1 - 2 | 779 | 79 | ||
| XM_008759889.1 | XP_008758111.1 | 1 - 2 | 781 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Edrf1
Genomic Map