Assay Details
Gene Symbol: LOC100360606 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002728842.4 | XP_002728888.3 | 3 - 4 | 1423 | 70 | |
XM_006230445.2 | XP_006230507.1 | 4 - 5 | 1588 | 70 | ||
XM_008759959.1 | XP_008758181.1 | 3 - 4 | 1675 | 70 | ||
XM_008759960.1 | XP_008758182.1 | 4 - 5 | 1320 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
