Assay Details
Gene Symbol: Mgmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012861.1 | NP_036993.1 | 3 - 4 | 357 | 102 | |
GenBank mRNA | M76704.1 | - | 2 - 3 | 302 | 102 | |
S61804.1 | - | 3 - 4 | 357 | 102 | ||
X54862.1 | - | 3 - 4 | 383 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
