Assay Details
| Gene Symbol: Oraov1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107565.1 | NP_001101035.1 | 2 - 3 | 524 | 71 | |
| XM_006230782.1 | XP_006230844.1 | 3 - 4 | 331 | 71 | ||
| XM_006230783.1 | XP_006230845.1 | 3 - 4 | 331 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Oraov1
Genomic Map