Assay Details
Gene Symbol: Ptpn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019253.3 | NP_062126.3 | 7 - 8 | 1368 | 69 | |
XM_006229252.2 | XP_006229314.1 | 12 - 13 | 2273 | 69 | ||
XM_006229253.2 | XP_006229315.1 | 12 - 13 | 2214 | 69 | ||
XM_006229254.2 | XP_006229316.1 | 11 - 12 | 2113 | 69 | ||
XM_006229255.2 | XP_006229317.1 | 11 - 12 | 2189 | 69 | ||
XM_006229257.2 | XP_006229319.1 | 10 - 11 | 2020 | 69 | ||
XM_008759346.1 | XP_008757568.1 | 12 - 13 | 1748 | 69 | ||
GenBank mRNA | BC092196.1 | - | 5 - 6 | 733 | 7313460 | 69 |
FQ211673.1 | - | 12 - 13 | 1877 | 69 | ||
FQ213308.1 | - | 9 - 10 | 1182 | 69 | ||
S49400.1 | - | 7 - 8 | 1368 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
