Assay Details
Gene Symbol: Prrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108482.1 | NP_001101952.1 | 1 - 2 | 128 | 118 | |
XM_006229098.2 | XP_006229160.1 | 1 - 2 | 786 | 118 | ||
XM_008759396.1 | XP_008757618.1 | 2 - 3 | 216 | 118 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001108482.1, XM_006229098.2, XM_008759396.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
