Assay Details
| Gene Symbol: Tsc22d4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006249186.2 | XP_006249248.1 | 5 - 6 | 2048 | 129 | |
| GenBank mRNA | BC079190.1 | - | 2 - 3 | 564 | 129 | |
| Gene Symbol: LOC100362783 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271236.1 | NP_001258165.1 | 563 | 129 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tsc22d4
Genomic Map