Assay Details
| Gene Symbol: Pla2g1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031585.1 | NP_113773.1 | 2 - 3 | 210 | 58 | |
| XM_008769222.1 | XP_008767444.1 | 2 - 3 | 318 | 58 | ||
| GenBank mRNA | D00036.1 | - | 2 - 3 | 210 | 58 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pla2g1b
Genomic Map