Assay Details
| Gene Symbol: Gpn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201991.1 | NP_973720.1 | 4 - 5 | 500 | 68 | |
| GenBank mRNA | AY513752.1 | - | 4 - 5 | 500 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpn3
Genomic Map