Assay Details
Gene Symbol: Ddx54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191548.1 | NP_001178477.1 | 10 - 11 | 1116 | 57 | |
XM_006249397.1 | XP_006249459.1 | 10 - 11 | 1192 | 57 | ||
XM_006249398.1 | XP_006249460.1 | 10 - 11 | 1192 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
