Assay Details
Gene Symbol: Sept14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001137670.1 | NP_001131142.1 | 7 - 8 | 852 | 74 | |
XM_008769202.1 | XP_008767424.1 | 7 - 8 | 949 | 74 | ||
XM_008769203.1 | XP_008767425.1 | 7 - 8 | 938 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
