Assay Details
Gene Symbol: Psmd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130430.1 | NP_569114.1 | 1 - 2 | 633 | 116 | |
GenBank mRNA | AF067728.1 | - | 1 - 2 | 633 | 116 | |
FQ214178.1 | - | 177 | 116 | |||
FQ225583.1 | - | 166 | 116 | |||
FQ227319.1 | - | 166 | 116 | |||
FQ229304.1 | - | 199 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
