Assay Details
| Gene Symbol: Mlec | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013983.1 | NP_001014005.1 | 3 - 4 | 721 | 117 | |
| GenBank mRNA | BC089839.1 | - | 3 - 4 | 721 | 7365814 | 117 |
More Information
Updated Mapping Information
Set Membership: |
|

Mlec
Genomic Map