Assay Details
| Gene Symbol: Wsb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007616.1 | NP_001007617.1 | 5 - 6 | 728 | 83 | |
| XM_008769220.1 | XP_008767442.1 | 4 - 5 | 665 | 83 | ||
| GenBank mRNA | BC081901.1 | - | 5 - 6 | 728 | 7110808 | 83 |
More Information
Updated Mapping Information
Set Membership: |
|

Wsb2
Genomic Map