Assay Details
| Gene Symbol: Rpl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053971.3 | NP_446423.2 | 2 - 3 | 366 | 55 | |
| XM_006249377.1 | XP_006249439.1 | 3 - 4 | 385 | 55 | ||
| XM_006249378.2 | XP_006249440.1 | 3 - 4 | 419 | 55 | ||
| GenBank mRNA | BC061784.1 | - | 2 - 3 | 362 | 55 | |
| BC078761.1 | - | 2 - 3 | 369 | 7123086 | 55 | |
| BC097295.1 | - | 2 - 3 | 370 | 7446680 | 55 | |
| FQ209419.1 | - | 1 - 2 | 44 | 55 | ||
| FQ213055.1 | - | 1 - 2 | 44 | 55 | ||
| FQ213078.1 | - | 1 - 2 | 44 | 55 | ||
| FQ216758.1 | - | 1 - 2 | 44 | 55 | ||
| FQ217744.1 | - | 1 - 2 | 44 | 55 | ||
| FQ219956.1 | - | 1 - 2 | 44 | 55 | ||
| FQ219982.1 | - | 1 - 2 | 44 | 55 | ||
| FQ220180.1 | - | 1 - 2 | 44 | 55 | ||
| FQ220346.1 | - | 1 - 2 | 44 | 55 | ||
| FQ220362.1 | - | 1 - 2 | 44 | 55 | ||
| FQ220471.1 | - | 1 - 2 | 44 | 55 | ||
| FQ220678.1 | - | 1 - 2 | 44 | 55 | ||
| FQ220699.1 | - | 1 - 2 | 44 | 55 | ||
| FQ220887.1 | - | 1 - 2 | 44 | 55 | ||
| FQ220959.1 | - | 1 - 2 | 44 | 55 | ||
| FQ221615.1 | - | 1 - 2 | 44 | 55 | ||
| FQ222080.1 | - | 1 - 2 | 44 | 55 | ||
| FQ222331.1 | - | 1 - 2 | 44 | 55 | ||
| FQ222465.1 | - | 1 - 2 | 44 | 55 | ||
| FQ223301.1 | - | 1 - 2 | 44 | 55 | ||
| FQ226632.1 | - | 1 - 2 | 44 | 55 | ||
| FQ227060.1 | - | 1 - 2 | 44 | 55 | ||
| FQ227100.1 | - | 1 - 2 | 44 | 55 | ||
| FQ227955.1 | - | 1 - 2 | 44 | 55 | ||
| FQ227974.1 | - | 1 - 2 | 44 | 55 | ||
| FQ228583.1 | - | 1 - 2 | 44 | 55 | ||
| FQ228665.1 | - | 1 - 2 | 44 | 55 | ||
| FQ229345.1 | - | 1 - 2 | 44 | 55 | ||
| FQ229355.1 | - | 1 - 2 | 44 | 55 | ||
| FQ229484.1 | - | 3 - 4 | 384 | 55 | ||
| FQ229571.1 | - | 1 - 2 | 44 | 55 | ||
| FQ229624.1 | - | 1 - 2 | 44 | 55 | ||
| FQ229675.1 | - | 1 - 2 | 44 | 55 | ||
| FQ229820.1 | - | 1 - 2 | 44 | 55 | ||
| FQ229854.1 | - | 1 - 2 | 44 | 55 | ||
| FQ229943.1 | - | 3 - 4 | 383 | 55 | ||
| FQ230171.1 | - | 1 - 2 | 44 | 55 | ||
| FQ232778.1 | - | 1 - 2 | 44 | 55 | ||
| FQ233475.1 | - | 1 - 2 | 44 | 55 | ||
| FQ234880.1 | - | 3 - 4 | 383 | 55 | ||
| FQ235095.1 | - | 1 - 2 | 44 | 55 | ||
| X87107.1 | - | 3 - 4 | 380 | 55 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpl6
Genomic Map