Assay Details
| Gene Symbol: Cmklr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022218.2 | NP_071554.2 | 1 - 2 | 95 | 69 | |
| XM_006249505.2 | XP_006249567.1 | 2 - 3 | 468 | 69 | ||
| XM_006249506.2 | XP_006249568.1 | 2 - 3 | 147 | 69 | ||
| XM_006249507.2 | XP_006249569.1 | 2 - 3 | 794 | 69 | ||
| XM_006249508.2 | XP_006249570.1 | 2 - 3 | 411 | 69 | ||
| XM_006249509.2 | XP_006249571.1 | 2 - 3 | 311 | 69 | ||
| GenBank mRNA | CB707803.1 | - | 2 - 3 | 351 | 69 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_022218.2,XM_006249505.2,XM_006249506.2,XM_006249507.2,XM_006249508.2,XM_006249509.2 transcripts.
Set Membership: |
|

Cmklr1
Genomic Map