Assay Details
| Gene Symbol: Ccdc92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083898.1 | NP_001077367.1 | 2 - 3 | 261 | 92 | |
| XM_006249292.2 | XP_006249354.1 | 2 - 3 | 518 | 92 | ||
| GenBank mRNA | CB796650.1 | - | 2 - 3 | 261 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc92
Genomic Map