Assay Details
| Gene Symbol: Ccdc92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083898.1 | NP_001077367.1 | 1 - 2 | 117 | 121 | |
| GenBank mRNA | CB796650.1 | - | 1 - 2 | 117 | 121 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001083898.1 transcripts.
Set Membership: |
|

Ccdc92
Genomic Map