Assay Details
| Gene Symbol: Ccl26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109488.1 | NP_001102958.1 | 1 - 2 | 135 | 58 | |
| XM_006249188.2 | XP_006249250.1 | 2 - 3 | 459 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccl26
Genomic Map