Assay Details
| Gene Symbol: Arpc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105933.1 | NP_001099403.1 | 3 - 4 | 290 | 110 | |
| GenBank mRNA | BC166554.1 | - | 3 - 4 | 249 | 7311069 | 110 |
| FQ219862.1 | - | 3 - 4 | 264 | 110 | ||
| FQ221109.1 | - | 3 - 4 | 250 | 110 | ||
| FQ232695.1 | - | 3 - 4 | 241 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arpc3
Genomic Map