Assay Details
| Gene Symbol: Arpc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105933.1 | NP_001099403.1 | 1 - 2 | 113 | 92 | |
| GenBank mRNA | BC166554.1 | - | 1 - 2 | 72 | 7311069 | 92 |
| FQ219862.1 | - | 1 - 2 | 87 | 92 | ||
| FQ221109.1 | - | 1 - 2 | 73 | 92 | ||
| FQ232695.1 | - | 1 - 2 | 64 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC166554.1, NM_001105933.1 transcripts.
Set Membership: |
|

Arpc3
Genomic Map