Assay Details
| Gene Symbol: Carkd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108402.1 | NP_001101872.1 | 3 - 4 | 262 | 142 | |
| XM_006253465.2 | XP_006253527.1 | 3 - 4 | 389 | 142 | ||
| XM_008771403.1 | XP_008769625.1 | 3 - 4 | 280 | 142 | ||
| GenBank mRNA | FQ225763.1 | - | 224 | 142 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Carkd
Genomic Map