Assay Details
Gene Symbol: Pcid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169144.1 | NP_001162615.1 | 6 - 7 | 381 | 60 | |
GenBank mRNA | BC160879.1 | - | 6 - 7 | 365 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
