Assay Details
Gene Symbol: Cars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177684.1 | NP_001171155.1 | 5 - 6 | 560 | 60 | |
XM_006253460.2 | XP_006253522.1 | 5 - 6 | 1008 | 60 | ||
XM_006253461.2 | XP_006253523.1 | 5 - 6 | 1006 | 60 | ||
XM_006253462.2 | XP_006253524.1 | 5 - 6 | 999 | 60 | ||
GenBank mRNA | BC166536.1 | - | 5 - 6 | 560 | 7191115 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
