Assay Details
| Gene Symbol: Cul4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127301.1 | NP_001120773.1 | 9 - 10 | 933 | 94 | |
| XM_008771401.1 | XP_008769623.1 | 8 - 9 | 1821 | 94 | ||
| GenBank mRNA | BC166799.1 | - | 9 - 10 | 933 | 7938235 | 94 |
| FQ210009.1 | - | 9 - 10 | 932 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cul4a
Genomic Map