Assay Details
| Gene Symbol: Mrps31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106091.1 | NP_001099561.1 | 2 - 3 | 455 | 59 | |
| XM_006253349.2 | XP_006253411.1 | 1676 | 59 | |||
| GenBank mRNA | BC158691.1 | - | 2 - 3 | 441 | 9021908 | 59 |
More Information
Updated Mapping Information
Set Membership: |
|

Mrps31
Genomic Map