Assay Details
| Gene Symbol: Grk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031096.1 | NP_112358.1 | 2 - 3 | 1010 | 128 | |
| GenBank mRNA | U63971.1 | - | 2 - 3 | 1010 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|

Grk1
Genomic Map