Assay Details
| Gene Symbol: Tnfsf13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109112.1 | NP_001102582.1 | 1 - 2 | 672 | 132 | |
| XM_006253480.2 | XP_006253542.1 | 1 - 2 | 996 | 132 | ||
| XM_006253481.2 | XP_006253543.1 | 1 - 2 | 996 | 132 | ||
| XM_006253482.2 | XP_006253544.1 | 1 - 2 | 995 | 132 | ||
| XM_006253483.2 | XP_006253545.1 | 1 - 2 | 994 | 132 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tnfsf13b
Genomic Map