Assay Details
| Gene Symbol: Np4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173299.2 | NP_775421.1 | 1 - 2 | 60 | 78 | |
| GenBank mRNA | U16684.1 | - | 1 - 2 | 61 | 78 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_173299.2,U16684.1 transcripts.
Set Membership: |
|

Np4
Genomic Map