Assay Details
Gene Symbol: RGD1304810 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107314.1 | NP_001100784.1 | 3 - 4 | 746 | 168 | |
XM_006253222.1 | XP_006253284.1 | 3 - 4 | 578 | 168 | ||
XM_006253223.2 | XP_006253285.1 | 3 - 4 | 2320 | 168 | ||
XM_008771300.1 | XP_008769522.1 | 2 - 3 | 677 | 168 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001107314.1, XM_006253222.1, XM_006253223.2, XM_008771300.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
