Assay Details
| Gene Symbol: RGD1304810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008771301.1 | XP_008769523.1 | 1 - 2 | 1047 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008771301.1 transcripts.
Set Membership: |
|

RGD1304810
Genomic Map