Assay Details
| Gene Symbol: Ppp1r3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138912.2 | NP_620267.2 | 1 - 2 | 208 | 106 | |
| GenBank mRNA | BC072514.1 | - | 1 - 2 | 208 | 7104413 | 106 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC072514.1, NM_138912.2 transcripts.
Set Membership: |
|

Ppp1r3b
Genomic Map