Assay Details
| Gene Symbol: Col4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135009.1 | NP_001128481.1 | 50 - 51 | 4957 | 69 | |
| GenBank mRNA | BC089096.1 | - | 4 - 5 | 538 | 69 | |
More Information
Updated Mapping Information
Set Membership: | 
		   
					
						
							 | 
				

Col4a1
Genomic Map