Assay Details
Gene Symbol: LOC290876 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008771386.1 | XP_008769608.1 | 3 - 4 | 814 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC290876 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008771386.1 | XP_008769608.1 | 3 - 4 | 814 | 107 |
Set Membership: |
![]() ![]() ![]() ![]() |