Assay Details
Gene Symbol: LOC290876 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037182.1 | NP_001032259.1 | 5 - 6 | 593 | 95 | |
XM_006253430.2 | XP_006253492.1 | 5 - 6 | 918 | 95 | ||
XM_006253432.2 | XP_006253494.1 | 5 - 6 | 919 | 95 | ||
XM_006253433.2 | XP_006253495.1 | 5 - 6 | 713 | 95 | ||
XM_008771386.1 | XP_008769608.1 | 5 - 6 | 957 | 95 | ||
GenBank mRNA | BC085810.1 | - | 5 - 6 | 552 | 96 | |
BC100249.1 | - | 5 - 6 | 593 | 7135908 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
