Assay Details
Gene Symbol: LOC290876 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037182.1 | NP_001032259.1 | 3 - 4 | 488 | 110 | |
XM_006253430.2 | XP_006253492.1 | 3 - 4 | 813 | 110 | ||
XM_006253432.2 | XP_006253494.1 | 3 - 4 | 814 | 110 | ||
XM_006253433.2 | XP_006253495.1 | 3 - 4 | 608 | 110 | ||
XM_008771386.1 | XP_008769608.1 | 3 - 4 | 814 | 148 | ||
GenBank mRNA | BC085810.1 | - | 3 - 4 | 447 | 110 | |
BC100249.1 | - | 3 - 4 | 488 | 7135908 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
