Assay Details
| Gene Symbol: LOC290876 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037182.1 | NP_001032259.1 | 4 - 5 | 538 | 63 | |
| XM_006253430.2 | XP_006253492.1 | 4 - 5 | 863 | 63 | ||
| XM_006253432.2 | XP_006253494.1 | 4 - 5 | 864 | 63 | ||
| XM_006253433.2 | XP_006253495.1 | 4 - 5 | 658 | 63 | ||
| GenBank mRNA | BC085810.1 | - | 4 - 5 | 497 | 63 | |
| BC100249.1 | - | 4 - 5 | 538 | 7135908 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

LOC290876
Genomic Map