Assay Details
Gene Symbol: Whsc1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106090.1 | NP_001099560.1 | 5 - 6 | 1467 | 61 | |
XM_006253307.2 | XP_006253369.1 | 5 - 6 | 1584 | 61 | ||
XM_006253309.2 | XP_006253371.1 | 5 - 6 | 1584 | 61 | ||
XM_006253311.2 | XP_006253373.1 | 5 - 6 | 1584 | 61 | ||
XM_008771349.1 | XP_008769571.1 | 5 - 6 | 3455 | 61 | ||
XM_008771350.1 | XP_008769572.1 | 5 - 6 | 1583 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
