Assay Details
| Gene Symbol: C4bpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016995.2 | NP_058691.2 | 1 - 2 | 333 | 145 | |
| XM_006249705.2 | XP_006249767.1 | 2 - 3 | 389 | 145 | ||
| XM_008769423.1 | XP_008767645.1 | 2 - 3 | 206 | 145 | ||
| GenBank mRNA | BC088152.1 | - | 1 - 2 | 333 | 7322205 | 145 |
| FQ209636.1 | - | 1 - 2 | 344 | 145 | ||
| FQ218474.1 | - | 1 - 2 | 314 | 145 | ||
| Z50052.1 | - | 1 - 2 | 312 | 145 | ||
More Information
Updated Mapping Information
Set Membership: |
|

C4bpb
Genomic Map