Assay Details
Gene Symbol: Crb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107182.1 | NP_001100652.1 | 11 - 12 | 4130 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P075614 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889800 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430247 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701633 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727703 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295234 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24209 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361133 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624157 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847956 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777832 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783878 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124269 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301744 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671588 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37093 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112474 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903260 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933938 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367417 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841674 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16494 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273374 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924765 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580445 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814729 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840253 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916198 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863579 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351243 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563191 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10679 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602249 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307740 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900595 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50728 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700578 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614044 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693584 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813720 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936035 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219349 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706569 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301149 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902558 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46105 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955935 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116679 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299393 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965193 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110861 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667539 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634166 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876365 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338510 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384288 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197157 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397712 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6570 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875091 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865551 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882028 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804940 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833301 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338919 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624620 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441082 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720079 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961236 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347781 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554932 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77510 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173899 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170670 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28852 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41157 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89298 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105593 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151256 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89739 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154934 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10700 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145918 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P075614 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P075614 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P075614 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P490600 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 71003 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 7404 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 16847 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 35167 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10768213 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10768216 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10768223 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10768217 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10768221 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10768219 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10768211 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10768214 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10768224 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10768215 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10768225 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10768210 | - | - |
Affymetrix Rat Clariom S Assay | PSR1300010481.rn.1 | TC1300001527.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1300010482.rn.1 | TC1300001527.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10768210 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22341834 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358883 | - | - |
